2-Bromo-6-methylpyridine

Warning

429112-10G 429112-1G
220066-644EA 157.23 USD
220066-644 220066-645
2-Bromo-6-methylpyridine
2-Bromo-6-methylpyridine
Formula: C₆H₆BrN
MDL Number: MFCD00040743
CAS Number: 5315-25-3
EINECS: 226-173-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR