Azobenzene ≥98%

Danger

424633-25G 424633-5G
220066-266EA 308 USD
220066-266 220066-267
Azobenzene ≥98%
Azobenzene
Formula: C₁₂H₁₀N₂
MW: 182.22 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 66…69 °C
Density: 1.203 g/cm³ (20 °C)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00003022
CAS Number: 103-33-3
EINECS: 203-102-5
UN: 3077
ADR: 9,III
Merck Index: 12,00948
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a5f45b1d83992325eab6ac608a29b7e4cc40073&vid=aded3940b8e2eae1f8b02939cc835c72f7c4a4da&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR