Sanger's reagent, purum p.a.

Warning

42085-50G 42085-250G
220065-944EA 149.69 USD
220065-944 220065-943
Sanger's reagent, purum p.a.
Sanger's reagent
Formula: C₆H₃FN₂O₄
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 24…26 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007056
CAS Number: 70-34-8
EINECS: 200-734-3
UN: 2811
ADR: 6.1,III
Merck Index: 13,04199

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR