N,N-Dimethyl-p-toluidine ≥98.5% (by GC), catalyst grade

Danger

Synonyms: N,N,4-Trimethylaniline, N,N-Dimethyl-4-methylaniline, 1-(Dimethylamino)-4-methylbenzol, 4,N,N-Trimethylaniline, 4-(Dimethylamino)toluene, 4-Dimethylaminotoluene

41690-100ML 41690-500ML
220065-550EA 63.97 USD
220065-550 220065-551
N,N-Dimethyl-p-toluidine ≥98.5% (by GC), catalyst grade
N,N-Dimethyl-p-toluidine

Catalyst grade for peroxide polymerization

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: -15 °C
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00008316
CAS Number: 99-97-8
EINECS: 202-805-4
UN: 1708
ADR: 6.1,II

Order Now

Specification Test Results

APPEARANCE (COLOR) Colorless to Dark Yellow and Colorless to Dark Green-Yellow
APPEARANCE (FORM) Liquid
PURITY (GC AREA %) ≥ 98.5 %
DENSITY D20/4 0.930 - 0.945
REFRACTIVE INDEX N20/D 1.546 - 1.548
PROTON NMR SPECTRUM CONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD 10 MONTHS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR