Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one absolute

Danger

Synonyms: Dimethylpropyleneurea, Dimethyl propylene urea, 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 1,3-Dimethyltetrahydropyrimidin-2(1H)-one, DMPU, 1,3-Dimethyltetrahydro-2(1H)-pyrimidinone, N,N'-Dimethyl propylene urea

41661-250ML 41661-1L
220065-527EA 236.92 USD
220065-527 220065-526
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one absolute
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one
Formula: C₆H₁₂N₂O
MW: 128.17 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00006550
CAS Number: 7226-23-5
EINECS: 230-625-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR