1-Chlorobutane, anhydrous ≥99.5%

Danger

Synonyms: n-Butyl Chloride

414255-1L 414255-100ML 414255-2L
220065-377EA 56.62 USD
220065-377 220065-376 220065-378
1-Chlorobutane, anhydrous ≥99.5%
1-Chlorobutane
Formula: CH₃(CH₂)₃Cl
MW: 92.57 g/mol
Boiling Pt: 78.4 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.8862 g/cm³ (20 °C)
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3,II
Merck Index: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR