1-Bromo-3,3-dimethylbutanone ≥97%

Warning

Synonyms: 1-Bromopinacolone, 1-Bromo-3,3-dimethyl-2-butanone

414131-25ML 414131-5ML
220065-363EA 179.85 USD
220065-363 220065-364
1-Bromo-3,3-dimethylbutanone ≥97%
1-Bromo-3,3-dimethylbutanone
Formula: C₆H₁₁BrO
MW: 179.06 g/mol
Boiling Pt: 47…49 °C (1 mmHg)
Melting Pt: 76 °C
Density: 1.3508 g/cm³ (0 °C)
Storage Temperature: Freezer
MDL Number: MFCD00000206
CAS Number: 5469-26-1
EINECS: 226-794-0
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR