Scandium(III) chloride, anhydrous

409359-5G 409359-1G
220063-661EA 1406.15 USD
220063-661 220063-660
Scandium(III) chloride, anhydrous
Scandium(III) chloride
Formula: Cl₃Sc
MW: 151.31 g/mol
Melting Pt: 940 °C
Density: 1.67 g/cm³ (940 °C)
MDL Number: MFCD00011221
CAS Number: 10361-84-9
EINECS: 233-799-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR