3-(Trimethylsilyl)propiolic acid ≥98%

Synonyms: 3-(Trimethylsilyl)propynoic acid

403482-5G 403482-1G
220064-466EA 216.46 USD
220064-466 220064-465
3-(Trimethylsilyl)propiolic acid ≥98%
3-(Trimethylsilyl)propiolic acid
Formula: C₆H₁₀O₂Si
MW: 142.23 g/mol
MDL Number: MFCD00190216
CAS Number: 5683-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR