N,N-Dimethylformamide di-tert-butyl acetal, Technical Grade

Synonyms: 1,1-Bis(1,1-dimethylethoxy)-N,N,N-trimethylamine, 1,1-Di-tert-butoxy-N,N-dimethylmethanamine, 1,1-Di-tert-butoxytrimethylamine

40263-10ML-F 40263-50ML-F
220064-360EA 420 USD
220064-360 220064-361
N,N-Dimethylformamide di-tert-butyl acetal, Technical Grade
N,N-Dimethylformamide di-tert-butyl acetal
Formula: C₁₁H₂₅NO₂
MW: 203.32 g/mol
Boiling Pt: 91…93 °C (1013 hPa)
Density: 0.85 g/cm³ (20 °C)
MDL Number: MFCD00015002
CAS Number: 36805-97-7
EINECS: 253-222-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR