N,N-Dimethylformamide diethyl acetal

Synonyms: 1,1-Diethoxy-N,N-dimethylmethanamine, 1,1-Diethoxytrimethylamine

40252-25ML-F 40252-100ML-F
220064-349EA 157.23 USD
220064-349 220064-348
N,N-Dimethylformamide diethyl acetal
N,N-Dimethylformamide diethyl acetal
Formula: C₇H₁₇NO₂
MW: 147.22 g/mol
Boiling Pt: 130 °C (1013 hPa)
Density: 0.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009227
CAS Number: 1188-33-6
EINECS: 214-707-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR