Benzene, anhydrous ≥99.8%

Danger

401765-6X1L 401765-1L 401765-2L 401765-100ML
220063-600EA 182.15 USD
220063-600 220063-598 220063-599 220064-298
Benzene, anhydrous ≥99.8%
Benzene
Formula: C₆H₆
Boiling Pt: 80.1 °C (1013 hPa)
Melting Pt: 5.5 °C
Density: 0.878 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3,II
REACH: 01-2119447106-44
Merck Index: 13,01066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c0146712ab33142aeeb3843b02381e6bda4028&vid=866487d2506bad13295f27385ab4dfe3176e9bb3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR