Iron(II) bromide

Synonyms: Iron dibromide, Ferrous bromide, Iron(II) bromide

400831-10G 400831-50G
220061-968EA 59.4 USD
220061-968 220061-969
Iron(II) bromide
Iron(II) bromide
Formula: Br₂Fe
MW: 215.66 g/mol
MDL Number: MFCD00016081
CAS Number: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR