Sodium tellurite(IV) ≥99% −100 mesh, Sigma-Aldrich®

Danger

Synonyms: Sodium tellurite, Disodium trioxotellurate

400688-10G 400688-50G
220061-959EA 200 USD
220061-959 220061-960
Sodium tellurite(IV) ≥99% −100 mesh, Sigma-Aldrich®
Sodium tellurite(IV)
Formula: Na₂O₃Te
MW: 221.58 g/mol
Melting Pt: 710 °C
MDL Number: MFCD00036141
CAS Number: 10102-20-2
EINECS: 233-268-4
UN: 3284
ADR: 6.1,II
Merck Index: 12,08834

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR