N,N-Dimethylformamide di-tert-butyl acetal ≥90.0% for GC derivatization

Warning

Synonyms: 1,1-Bis(1,1-dimethylethoxy)-N,N,N-trimethylamine, 1,1-Di-tert-butoxy-N,N-dimethylmethanamine, 1,1-Di-tert-butoxytrimethylamine

395005-10X1ML 395005-5ML 395005-25ML
220063-490EA 232.31 USD
220063-490 220063-492 220063-491
N,N-Dimethylformamide di-tert-butyl acetal ≥90.0% for GC derivatization
N,N-Dimethylformamide di-tert-butyl acetal
Formula: C₁₁H₂₅NO₂
MW: 203.32 g/mol
Boiling Pt: 91 - 93 °C (1013 hPa)
Density: 0.85 g/cm³ (20 °C)
MDL Number: MFCD00015002
CAS Number: 36805-97-7
EINECS: 253-222-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=039d4b17a9cd3649f9f3329b343a8cf1d0ac09d3&vid=1789c0633d842d064e548947b35289cfa5e17bc1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

APPEARANCE (COLOR) Colorless to Yellow
APPEARANCE (FORM) Liquid
PURITY (GC AREA %) ≥ 90.0 %
PROTON NMR SPECTRUM CONFORMS TO STRUCTURE

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR