N,N-Dimethylformamide dipropyl acetal ≥97.5% for GC derivatization

Warning

Synonyms: 1,1-Dipropoxytrimethylamine, N,n-dimethylformamidedipropylacetal, 1,1-Dipropoxy-N,N-dimethylmethylamine

394998-10X1ML 394998-25ML
220063-487EA 153.14 USD
220063-487 220063-488
N,N-Dimethylformamide dipropyl acetal ≥97.5% for GC derivatization
N,N-Dimethylformamide dipropyl acetal

N,N-Dimethylformamide dipropyl acetal is an derivatizing reagent.

Formula: C₉H₂₁NO₂
MW: 175.27 g/mol
Boiling Pt: 67 °C (1013 hPa)
Density: 0.8537 g/cm³ (20 °C)
MDL Number: MFCD00009374
CAS Number: 6006-65-1
EINECS: 227-855-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60fbf35b63f3859752813a2b4f7edd6f1d5d61b2&vid=f4253d577dcb230682f45345833119cff95ebda9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

APPEARANCE (COLOR) Colorless
APPEARANCE (FORM) Liquid
PURITY (GC AREA %) ≥ 97.5 %
INFRARED SPECTRUM CONFORMS TO STRUCTURE

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR