Perylene, sublimated

394475-1G 394475-5G
220063-591EA 125.54 USD
220063-591 220063-592
Perylene, sublimated
Perylene
Formula: C₂₀H₁₂
MW: 252.32 g/mol
Boiling Pt: 443 °C (1013 hPa)
Melting Pt: 273…278 °C
Density: 1.362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004142
CAS Number: 198-55-0
EINECS: 205-900-9
Merck Index: 12,07325

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR