1-Benzyl-3-methylimidazolium hexafluorophosphate

39447-50G-F 39447-5G-F
220063-589EA 91.85 USD
220063-589 220063-590
1-Benzyl-3-methylimidazolium hexafluorophosphate
1-Benzyl-3-methylimidazolium hexafluorophosphate
Formula: C₁₁H₁₃F₆N₂P
MW: 318.2 g/mol
MDL Number: MFCD07784448
CAS Number: 433337-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR