Potassium permanganate concentrate for dilution to 0.1 N, Fixanal
|
|
Synonyms:
Permanganic acid potassium salt
38130-6X1EA-R
38130-1EA-R
220061-666EA
59.68
USD
220061-666
220061-665
Potassium permanganate concentrate for dilution to 0.1 N, Fixanal
Potassium permanganate
1/50 MOL POTASSIUM PERMANGANATE FIXANAL (3.161G KMNO4 IN 1EA)
Formula:
KMnO₄ MW: 158.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011364 CAS Number: 7722-64-7 EINECS: 231-760-3 UN: 1490 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...