Antimony(III) fluoride ≥99.8%

Danger

Synonyms: Antimony trifluoride

381292-500G 381292-100G
220061-664EA 684.62 USD
220061-664 220061-663
Antimony(III) fluoride ≥99.8%
Antimony(III) fluoride
Formula: F₃Sb
MW: 178.76 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 292 °C
Density: 4.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011218
CAS Number: 7783-56-4
EINECS: 232-009-2
UN: 2923
ADR: 8,II
Merck Index: 13,00715

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR