Dithiodiglycol (bis(2-hydroxyethyl) disulfide), Technical Grade

Synonyms: 2-Hydroxyethyl disulphide, 2,2'-Dithiobisethanol, 2,2'-Dithiodiethanol, Bis(2-hydroxyethyl) disulphide

380474-250ML 380474-50ML
220062-725EA 383.15 USD
220062-725 220062-726
Dithiodiglycol (bis(2-hydroxyethyl) disulfide), Technical Grade
Dithiodiglycol (bis(2-hydroxyethyl) disulphide)
Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 27.5 °C
Density: 1.27 g/cm³ (20 °C)
MDL Number: MFCD00002906
CAS Number: 1892-29-1
EINECS: 217-576-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR