4-Chloro-3-nitrobenzonitrile ≥97%

380059-5G 380059-25G
220062-700EA 48.46 USD
220062-700 220062-699
4-Chloro-3-nitrobenzonitrile ≥97%
4-Chloro-3-nitrobenzonitrile
Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00016987
CAS Number: 939-80-0
EINECS: 213-364-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR