(S)-(+)-2-Hydroxyisovaleric acid ≥99%

Synonyms: (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid

379093-5G 379093-1G
220062-600EA 234.77 USD
220062-600 220062-599
(S)-(+)-2-Hydroxyisovaleric acid ≥99%
(S)-(+)-2-Hydroxyisovaleric acid
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
MDL Number: MFCD00066443
CAS Number: 17407-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR