Sodium hydrogen carbonate-[¹³C]

Synonyms: (13C)-Sodium bicarbonate

372382-1G 372382-250MG
220061-777EA 226.15 USD
220061-777 220061-778
Sodium hydrogen carbonate-[¹³C]
Sodium hydrogen carbonate-[¹³C]
Formula: NaHCO₃
MW: 85 g/mol
CAS Number: 87081-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR