Ammonium acetate ≥99.999%

Warning

Synonyms: Acetic acid ammonium salt

372331-10G 372331-100G
220061-773EA 128.15 USD
220061-773 220061-772
Ammonium acetate ≥99.999%
Ammonium acetate
Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR