1,1,1,3,5,5,5-Heptamethyltrisiloxane

Synonyms: Methylbis(trimethylsilyloxy)silane

370886-50ML 370886-250ML
220061-700EA 88.62 USD
220061-700 220061-699
1,1,1,3,5,5,5-Heptamethyltrisiloxane
1,1,1,3,5,5,5-Heptamethyltrisiloxane
Formula: C₇H₂₂O₂Si₃
MW: 222.51 g/mol
MDL Number: MFCD00048000
CAS Number: 1873-88-7
EINECS: 217-496-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR