Nickel(II) chloride hydrate

Danger

364304-100G 364304-500G
220059-166EA 110.15 USD
220059-166 220059-167
Nickel(II) chloride hydrate
Nickel(II) chloride hydrate
Formula: Cl₂Ni·H₂O
MW: 147.61 g/mol
MDL Number: MFCD00149808
CAS Number: 69098-15-3
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR