2-Butene cis- and trans-mixture ≥99%

Danger

363359-50G 363359-200G
220060-960EA 340.38 USD
220060-960 220060-959
2-Butene cis- and trans-mixture ≥99%
2-Butene
Formula: C₄H₈
MW: 56.11 g/mol
Boiling Pt: 0.88 °C (1013 hPa)
Melting Pt: –127 °C
Density: 0.6105 g/cm³ (18 °C)
CAS Number: 107-01-7
EINECS: 203-452-9
Merck Index: 12,01549

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR