Boron trifluoride dihydrate ≥96%

Danger

359963-100ML 359963-500ML
220060-600EA 48.88 USD
220060-600 220060-601
Boron trifluoride dihydrate ≥96%
Boron trifluoride dihydrate
Formula: BF₃·2H₂O
MW: 103.84 g/mol
Boiling Pt: 58.5…60 °C (1 mmHg)
Melting Pt: 6.2 °C
Density: 1.636 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149901
CAS Number: 13319-75-0
EINECS: 231-569-5
UN: 2851
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR