Potassium sorbate

Warning

Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

359769-1KG 359769-250G
220060-447EA 158.31 USD
220060-447 220060-448
Potassium sorbate
Potassium sorbate
Formula: CH₃CH=CHCH=CHCOOK
MW: 150.22 g/mol
Melting Pt: 270 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016546
CAS Number: 24634-61-5
EINECS: 246-376-1
Merck Index: 13,07756
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR