1-Butylimidazole

Synonyms: N-Butyl-1H-imidazole, 1-Butyl-1H-imidazole, N-Butylimidazole

348414-100G 348414-25G
220059-075EA 114.15 USD
220059-075 220059-076
1-Butylimidazole
1-Butylimidazole
Formula: C₇H₁₂N₂
MW: 124.19 g/mol
MDL Number: MFCD00042753
CAS Number: 4316-42-1
EINECS: 224-335-9
UN: 2690
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR