4-Amino-m-anisic acid ≥98%

Synonyms: 4-Amino-3-methoxybenzenecarboxylic acid

347027-5G 347027-1G
220059-376EA 161.54 USD
220059-376 220059-375
4-Amino-m-anisic acid ≥98%
4-Amino-m-anisic acid
Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00016539
CAS Number: 2486-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR