(1R,2R)-(-)-1,2-Diaminocyclohexane ≥98%

Danger

Synonyms: (R,R)-DACH, (1R,2R)-Cyclohexane-1,2-diamine

346721-1G 346721-5G
220059-050EA 22.62 USD
220059-050 220059-051
(1R,2R)-(-)-1,2-Diaminocyclohexane ≥98%
(1R,2R)-(-)-1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: 14…15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062985
CAS Number: 20439-47-8
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR