2-Chloro-4-hydroxybenzonitrile ≥98%

346071-10G 346071-5G
220059-284EA 350 USD
220059-284 220059-285
2-Chloro-4-hydroxybenzonitrile ≥98%
2-Chloro-4-hydroxybenzonitrile
Formula: C₇H₄ClNO
MW: 153.57 g/mol
MDL Number: MFCD00052185
CAS Number: 3336-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR