Carbon disulfide ≥99.9%, ReagentPlus®

Danger

342270-100ML 342270-1L
220058-728EA 42.77 USD
220058-728 220058-729
Carbon disulfide ≥99.9%, ReagentPlus®
Carbon disulphide
Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46.2 °C (1013 hPa)
Melting Pt: -111.6 °C
Density: 1.2632 g/cm³ (20 °C)
Flash Pt: -30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR