2,3-Diaminophenol ≥97%

Synonyms: 3-Hydroxy-o-phenylenediamine

340618-25G 340618-1G 340618-5G
220057-127EA 81.69 USD
220057-127 220057-126 220057-128
2,3-Diaminophenol ≥97%
2,3-Diaminophenol
Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD00075199
CAS Number: 59649-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR