Allyl bromide ≥97%, Reagent Grade

Synonyms: 3-Bromopropene, 3-Propenyl bromide

337528-250ML 337528-1L
220057-801EA 60.2 USD
220057-801 220057-800
Allyl bromide ≥97%, Reagent Grade
Allyl bromide
Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 70 °C (1013 hPa)
Melting Pt: -119 °C
Density: 1.4 g/cm³ (20 °C)
Flash Pt: -1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000244
CAS Number: 106-95-6
EINECS: 203-446-6
UN: 1099
ADR: 3,I
Merck Index: 13,00285

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR