Thallium(I) bromide, anhydrous ≥99.999%

Synonyms: Thallium bromide

336270-10G 336270-50G
220057-858EA 178.46 USD
220057-858 220057-859
Thallium(I) bromide, anhydrous ≥99.999%
Thallium(I) bromide
Formula: BrTl
MW: 284.29 g/mol
Boiling Pt: 815 °C (1013 hPa)
Melting Pt: 460 °C
Density: 7.56 g/cm³ (20 °C)
MDL Number: MFCD00011273
CAS Number: 7789-40-4
EINECS: 232-163-0
UN: 1707
ADR: 6.1,II
Merck Index: 12,09393

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR