Sodium stannate trihydrate ≥95%

Warning

336262-500G 336262-100G
220057-856EA 184.62 USD
220057-856 220057-855
Sodium stannate trihydrate ≥95%
Sodium stannate trihydrate
Formula: Na₂O₃Sn·3H₂O
MDL Number: MFCD00150189
CAS Number: 12209-98-2
EINECS: 235-030-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR