3-Amino-4-methylbenzyl alcohol ≥97%

Synonyms: 3-Amino-4-methylbenzyl Alcohol

335320-5G 335320-25G
220057-035EA 57.08 USD
220057-035 220057-440
3-Amino-4-methylbenzyl alcohol ≥97%
3-Amino-4-methylbenzyl alcohol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00075051
CAS Number: 81863-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR