Indium(III) chloride ≥98%

Synonyms: Indium Trichloride Chloride, Indium trichloride

334065-10G 334065-50G
220057-559EA 170.77 USD
220057-559 220057-560
Indium(III) chloride ≥98%
Indium(III) chloride
Formula: Cl₃In
MW: 221.18 g/mol
Boiling Pt: 800 °C (1013 hPa)
Melting Pt: 586 °C
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011058
CAS Number: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8,II
Merck Index: 13,04980

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR