Triphosgene ≥98%, Reagent Grade

330752-25G 330752-100G 330752-5G
220057-079EA 158.31 USD
220057-079 220057-078 220057-080
Triphosgene ≥98%, Reagent Grade
Triphosgene
Formula: C₃Cl₆O₃
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 77…81 °C
Density: 1.629 g/cm³ (80 °C)
MDL Number: MFCD00062848
CAS Number: 32315-10-9
EINECS: 250-986-3
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR