trans-1,4-Diaminocyclohexane

Synonyms: trans-Hexahydro-1,4-phenylenediamine, trans-Cyclohexane-1,4-diamine, trans-1,4-Cyclohexanediamine

32851-100G 32851-25G
220056-619EA 221.85 USD
220056-619 220056-620
trans-1,4-Diaminocyclohexane
trans-1,4-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: 70 °C
MDL Number: MFCD00075174
CAS Number: 2615-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR