Dysprosium trichloride, anhydrous ≥99.99%

Warning

325546-5G 325546-25G
220055-845EA 424.62 USD
220055-845 220055-844
Dysprosium trichloride, anhydrous ≥99.99%
Dysprosium trichloride
Formula: Cl₃Dy
MW: 268.86 g/mol
Melting Pt: 642…652 °C
Density: 3.67 g/cm³ (0 °C)
MDL Number: MFCD00010984
CAS Number: 10025-74-8
EINECS: 233-039-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR