Selenium dioxide ≥98%, Reagent Grade

Danger

325473-100G 325473-5G 325473-500G
220055-830EA 65.55 USD
220055-830 220055-832 220055-831
Selenium dioxide ≥98%, Reagent Grade
Selenium dioxide
Formula: SeO₂
MW: 110.96 g/mol
Melting Pt: 315 °C
Density: 3.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003562
CAS Number: 7446-08-4
EINECS: 231-194-7
UN: 3283
ADR: 6.1,II
Merck Index: 13,08508

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR