trans-1,4-Dichloro-2-butene ≥98%

Synonyms: (E)-1,4-Dichloro-2-butene

324515-5G 324515-25G
220056-178EA 68.06 USD
220056-178 220056-177
trans-1,4-Dichloro-2-butene ≥98%
trans-1,4-Dichloro-2-butene
Formula: C₄H₆Cl₂
MW: 125 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.1873 g/cm³ (20 °C)
Storage Temperature: Refrigerator
CAS Number: 110-57-6
EINECS: 203-779-7
UN: 3390
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR