Ferroin (1,10-phenanthrolineferrous sulfate complex) 0.1% in aqueous solution indicator

Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate

318922-500ML 318922-100ML
220055-480EA 371 USD
220055-480 220055-479
Ferroin (1,10-phenanthrolineferrous sulfate complex) 0.1% in aqueous solution indicator
Ferroin (1,10-phenanthrolineferrous sulphate complex)
Formula: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
MDL Number: MFCD00036428
CAS Number: 14634-91-4
EINECS: 238-676-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR