Diallyl disulfide ≥80%, Technical Grade

Warning

Synonyms: Diallyldisulphide, Allyl disulphide

317691-100G 317691-25G
220052-724EA 159.38 USD
220052-724 220052-725
Diallyl disulfide ≥80%, Technical Grade
Diallyl disulphide
Formula: C₆H₁₀S₂
MW: 146.28 g/mol
Boiling Pt: 198 °C (1013 hPa)
Density: 1.0237 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008656
CAS Number: 2179-57-9
EINECS: 218-548-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR