Isophorone diisocyanate ( mixture of isomers) ≥98%

Danger

317624-250ML 317624-5ML 317624-1L
220055-434EA 117.55 USD
220055-434 220055-225 220055-433
Isophorone diisocyanate ( mixture of isomers) ≥98%
Isophorone diisocyanate
Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
MDL Number: MFCD00064956
CAS Number: 4098-71-9
EINECS: 223-861-6
UN: 2290
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR