Phosphorus(III) chloride ≥99.999%

Danger

310115-100G 310115-25G
220054-514EA 589.77 USD
220054-514 220054-515
Phosphorus(III) chloride ≥99.999%
Phosphorus(III) chloride
Formula: Cl₃P
Boiling Pt: 76 °C (1013 hPa)
Melting Pt: –112 °C
Density: 1.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011438
CAS Number: 7719-12-2
EINECS: 231-749-3
UN: 1809
ADR: 6.1,I
Merck Index: 12,07515

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR